missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) perchlorate hydrate, Reagent Grade

Product Code. 11301758
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11301758 100 g 100g
11391748 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11301758 Supplier Thermo Scientific Alfa Aesar Supplier No. 011646.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 335159-18-7 | Cl2FeO8 | 254.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335159-18-7
Molecular Formula Cl2FeO8
Molecular Weight (g/mol) 254.74
MDL Number MFCD00150234
InChI Key CKFMJXZQTNRXGX-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) diperchlorate
SMILES [Fe++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Melting Point >100°C (decomposition)
Physical Form Crystalline
Quantity 100 g
UN Number UN1481
Solubility Information Soluble in alcohol and water (?98g/100 mL H2O at 0?°C).
Formula Weight 254.75 (Anhydrous)
Grade Reagent
Sensitivity Hygroscopic
Chemical Name or Material Iron(II) perchlorate hydrate
compliance-icons compliance-icons
Product Identifier
  • Iron(II) perchlorate hydrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.