missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) perchlorate hydrate, Reagent Grade

Product Code. 11391748
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391748 500 g 500g
11301758 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391748 Supplier Thermo Scientific Alfa Aesar Supplier No. 011646.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 335159-18-7 | Cl2FeO8 | 254.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335159-18-7
Molecular Formula Cl2FeO8
Molecular Weight (g/mol) 254.74
MDL Number MFCD00150234
InChI Key CKFMJXZQTNRXGX-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) diperchlorate
SMILES [Fe++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Melting Point >100°C (decomposition)
Physical Form Crystalline
Quantity 500 g
UN Number UN1481
Solubility Information Soluble in alcohol and water (?98g/100 mL H2O at 0?°C).
Formula Weight 254.75 (Anhydrous)
Grade Reagent
Sensitivity Hygroscopic
Chemical Name or Material Iron(II) perchlorate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.