missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) Phthalocyanine, TRC

Product Code. 30391295
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30391295 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391295 Supplier Toronto Research Chemicals Supplier No. TRCI775260250MG
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iron(II) Phthalocyanine
CAS 132-16-1
Molecular Formula C32H16FeN8
Quantity 250 mg
Synonym 29H,31H-Phthalocyanine Iron Complex, (Phthalocyaninato)iron, (Phthalocyaninato)iron(II), Ferrophthalocyanine, Ferrophthalocyanine Complex, Ferrous phthalocyanine, Iron Phthalocyanine, Iron(2+) Phthalocyaninate, Iron(2+) Phthalocyanine, Iron(II) Phthalocyanine, P 0774, P 26, Phthalocyanine iron, Phthalocyanine-iron(II), Sanyo P 26, [Phthalocyanato(2-)]iron, [Phthalocyaninato(2-)]iron
InChI Formula InChI=1S/C32H16N8.Fe/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;/h1-16H;/q-2;+2
SMILES C1(/N=C2N=C(C3=CC=CC=C\23)/N=C(C4=CC=CC=C/54)\[N-]C5=N/C(C6=CC=CC=C/76)=NC7=N/8)=C(C=CC=C9)C9=C8[N-]1.[Fe+2]
IUPAC Name phthalocyanine-29,31-diide
Molecular Weight (g/mol) 568.37
Formula Weight 568.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.