missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) sulfate concentrate, Solstice

Product Code. 15675190
Change view
Click to view available options
Quantity:
1 Pc.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15675190 1 Pc. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15675190 Supplier Honeywell-Fluka Supplier No. 380471EA
  • ONLINE EXCLUSIVE PRICE
    91.00 €
    85.34 € / Pack of 1
    Save 5.66 €
    6% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for 1L standard solution, 0.1 M FeSO4

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 10028-21-4
Physical Form Crystalline Powder
Quantity 1 Pc.
MDL Number MFCD00011014
UN Number NONH for all modes of transport
Molecular Weight (g/mol) 278.01
Percent Purity 99.0 to 103.4%
Grade ACS
Chemical Name or Material Iron(II) sulfate concentrate
compliance-icons
Product Identifier
  • Iron(II) sulphate concentrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: iron (II) sulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.