missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) Sulfate Heptahydrate, 77-90%, Extra Pure, Dried, SLR, Fisher Chemical

Product Code. 10794561 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10794561 1 kg Plastic powder jar 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10794561 Supplier Fisher Chemical Supplier No. I/1159/60
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 300°C
Melting Point 64°C
CAS 7782-63-0
Color Blue-Green
pH 3 to 4
Physical Form Solid
Quantity 1 kg
Molecular Formula FeH14O11S
MDL Number MFCD00149719
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
IUPAC Name λ²-iron(2+) heptahydrate sulfate
Molecular Weight (g/mol) 278.01
PubChem CID 62662
ChEBI CHEBI:75836
Packaging Plastic powder jar
Vapor Pressure 14.6mmHg at 25°C
Show More Show Less
compliance-icons
Product Identifier
  • Iron (II) sulfate - dried
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.