missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) sulfate heptahydrate, 99.5%, for analysis

Product Code. 11994241 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
1kg
250g
5g
5kg
Product Code. Quantity Packaging unitSize
11994241 5 kg Plastic bottle 5kg
10752051 5 g Glass Bottle 5g
10782241 250 g Plastic bottle 250g
10286300 1 kg Plastic bottle 1kg
4 options
This item is not returnable. View return policy

Product Code. 11994241

Brand: Thermo Scientific Acros 201390051

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7782-63-0 | FeH14O11S | 278.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
PubChem CID 62662
ChEBI CHEBI:75836
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Melting Point 64.0°C
Color Blue to Green
Physical Form Crystals
Quantity 5 kg
Assay Percent Range 99.5%
Linear Formula FeSO4·7H2O
Fieser 01,393; 04,237; 05,308
Merck Index 15,4084
Solubility Information Solubility in water: 25.6 g/100mL (20°C). Other solubilities: insoluble in alcohol
IUPAC Name iron(2+);sulfate;heptahydrate
Formula Weight 278.01
Percent Purity 99.5%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Iron(II) sulfate heptahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Iron(II) sulfate heptahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Iron(II) sulfate heptahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.