missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Melting Point | 64°C |
| Color | Blue-Green |
| pH | 3 to 4 |
| Physical Form | Solid |
| Quantity | 1 kg |
| IUPAC Name | λ²-iron(2+) heptahydrate sulfate |
| Formula Weight | 278.01g/mol |
| Packaging | Plastic powder jar |
| Vapor Pressure | 14.6mmHg at 25°C |
Safety and Handling
Product Identifier
- Iron(II) sulfate heptahydrate
- Warning
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: Iron (II) Sulfate heptahydrate, Ferroussulfate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?