missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis)

Product Code. 11352058
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352058 5 g 5g
11342058 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352058 Supplier Thermo Scientific Alfa Aesar Supplier No. 012967.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7782-63-0 | FeH14O11S | 278.01 g/mol

Iron(II) sulfate heptahydrate is used as a precursor to prepare other iron compounds such as a lawn conditioner and a mordant for wool dyeing. It is actively used in the manufacture of ink including iron gall ink. As a reducing agent, it participates in the reduction of chromate in cement. It is also used in industrial water treatment plants to remove phosphate. It is also used in the gold refining process to precipitate metallic gold. Woodworkers use aqueous solutions of ferrous sulfate to color maple wood a silvery hue. It finds application in the treatment of iron chlorosis, which arises due to deficiency of iron in horticulture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
PubChem CID 62662
ChEBI CHEBI:75836
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Melting Point 64°C (in -6H2O)
Color Green
Physical Form Crystalline
Quantity 5 g
Linear Formula FeSO4·7H2O
Merck Index 14,4057
Solubility Information Soluble in water. Insoluble in alcohol.
IUPAC Name iron(2+);sulfate;heptahydrate
Formula Weight 278.02 (151.91 Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Sensitivity Air Sensitive
Density 1.897 g/mL
Assay (metals basis)
Chemical Name or Material Iron(II) sulfate heptahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Iron(II) sulfate heptahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.