missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) Sulfide, 99.9%, For Trace Metals Basis, Honeywell™

Product Code. 15604170 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15604170 10 g Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15604170 Supplier Honeywell Research Chemicals Supplier No. 34316110G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1317-37-9
Quantity 10 g
Molecular Formula FeS
MDL Number MFCD00011013
UN Number NONH for all modes of transport
Synonym ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
InChI Key GNVXPFBEZCSHQZ-UHFFFAOYSA-N
SMILES [S--].[Fe++]
IUPAC Name sulfanylideneiron
Molecular Weight (g/mol) 87.91
PubChem CID 14828
Formula Weight 87.91g/mol
Percent Purity 99.9% (trace metals basis)
Packaging Glass Bottle
Appearance Solid
Density 4.84g/mL at 25°C
Chemical Name or Material Iron(II) sulfide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.