missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) sulfide, fused sticks

Product Code. 10206580 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1kg; Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10206580 1kg; Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10206580 Supplier Thermo Scientific Acros Supplier No. 233300010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1317-37-9 | FeS | 87.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 1195.0°C
CAS 1317-37-9
Color Black-Gray
Physical Form Sticks
Quantity 1 kg
Molecular Formula FeS
Linear Formula FeS
MDL Number MFCD00011013
Merck Index 15, 4085
Synonym ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in acids with the evolution of h2s
InChI Key GNVXPFBEZCSHQZ-UHFFFAOYSA-N
SMILES [S--].[Fe++]
IUPAC Name sulfanylideneiron
Molecular Weight (g/mol) 87.91
PubChem CID 14828
Formula Weight 87.92
Percent Purity ∼25% (S)
Packaging Plastic bottle
Specific Gravity 4.84
Density 4.8400g/mL
Chemical Name or Material Iron(II) sulfide, c.p., 60-67% Fe
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.