missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) sulfide, tech.

Product Code. 11453037
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453037 5000 g 5000g
11443037 250 g 250g
11433037 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453037 Supplier Thermo Scientific Alfa Aesar Supplier No. A15569.0I
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1317-37-9 | FeS | 87.91 g/mol

Iron(II) sulfide is used as a pigment in hair dyes and ceramics, glasses and bottles, and paint, Also used in lubricants and anodes, as a hydrogen sulfide source, and to treat exhaust gases. And it is also used as resulphurizing agent in the production of carbon, alloy and stainless types of free cutting steels. It acts as an alloying agent to improve machine ability of steel castings which are used to manufacture various steel components.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(II) sulfide is used as a pigment in hair dyes and ceramics, glasses and bottles, and paint, Also used in lubricants and anodes, as a hydrogen sulfide source, and to treat exhaust gases. And it is also used as resulphurizing agent in the production of carbon, alloy and stainless types of free cutting steels. It acts as an alloying agent to improve machine ability of steel castings which are used to manufacture various steel components.

Solubility
Soluble in water(0.0062g/L ), in acids with evolution of hydrogen sulfide. Insoluble in nitric acid.

Notes
Moisture Sensitive. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agent, acids, air, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-37-9
Molecular Formula FeS
Molecular Weight (g/mol) 87.91
MDL Number MFCD00011013
InChI Key GNVXPFBEZCSHQZ-UHFFFAOYSA-N
Synonym ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
PubChem CID 14828
SMILES [S--].[Fe++]

Specifications

Melting Point 1195°C
Quantity 5000 g
Merck Index 14,4058
Solubility Information Soluble in water(0.0062g/L ),in acids with evolution of hydrogen sulfide. Insoluble in nitric acid.
IUPAC Name λ²-iron(2+) sulfanediide
Formula Weight 87.91
Grade Technical
Odor Odorless
Sensitivity Air and moisture sensitive
Density 4.74
Chemical Name or Material Iron(II) sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.