missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) Sulfide, Technical grade, Honeywell™ Fluka™

Product Code. 15652820 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Poly bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15652820 250 g Poly bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15652820 Supplier Honeywell-Fluka Supplier No. 268704250G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Technical Grade

  • Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
  • Variation in color and physical form is possible
TRUSTED_SUSTAINABILITY

Specifications

Melting Point >1190°C
CAS 1317-37-9
Color Brown
Physical Form Powder
Quantity 250 g
Molecular Formula FeS
MDL Number MFCD00011013
UN Number NONH for all modes of transport
Synonym ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
InChI Key GNVXPFBEZCSHQZ-UHFFFAOYSA-N
SMILES [S--].[Fe++]
IUPAC Name λ²-iron(2+) sulfanediide
Molecular Weight (g/mol) 87.91
PubChem CID 14828
Formula Weight 87.91g/mol
Grade Technical
Odor Odorless
Packaging Poly bottle
Density 4.84g/mL at 25°C
Chemical Name or Material Iron(Ii) Sulfide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.