missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) Sulfide, Technical, Fisher Chemical

Product Code. 10060210 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10060210 1 kg Plastic powder jar 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10060210 Supplier Fisher Chemical Supplier No. I/1170/60
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 1195°C
CAS 1317-37-9
Color Gray
Physical Form Solid
Quantity 1 kg
Molecular Formula FeS
MDL Number MFCD00011013
Synonym ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
InChI Key GNVXPFBEZCSHQZ-UHFFFAOYSA-N
SMILES [S--].[Fe++]
IUPAC Name λ²-iron(2+) sulfanediide
Molecular Weight (g/mol) 87.91
PubChem CID 14828
Formula Weight 87.92g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.