Learn More
Iron(III) 2,4-pentanedionate
Description
Iron(III) 2,4-pentanedionate is used as a precatalyst and reagent in organic chemistry. It acts as a precursor for highly crystalline (Zn, Fe)Fe2O4 films and magnetic property measurements of these films. It catalyzes the dimerization of isoprene to a mixture of 1,5-dimethyl-1,5-cyclooctadiene and 2,5-dimethyl-1,5-cyclooctadiene, ring-opening polymerization of 1,3-benzoxazine and catalyze the reaction of N-sulfonyl oxaziridines with olefins to form 1,3-oxazolidine products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∼183°C (decomposition) |
| Color | Red |
| Quantity | 100 g |
| Beilstein | 4157960 |
| Solubility Information | Soluble in toluene. |
| Formula Weight | 353.17 |
| Physical Form | Powder |
| Chemical Name or Material | Iron(III) 2,4-pentanedionate |
Safety and Handling
- Iron(III) 2,4-pentanedionate
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.