missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) bromide, anhydrous, 98+%, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 031125.14

Code : NEW

Additional Details : CAS Number : 10031-26-2 Weight : 0.02500kg


Product Code. 11367408

  • 185.00 € / 25g
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Iron(III) bromide is used as a Lewis acid catalyst in the halogenation of aromatic compounds. It is also used in the preparation of 2,5-dibromo-4-aryl-1,3-pentadiene by reacting with homoallenic bromohydrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(decomposition)
Solid
FeBr{3}
10031-26-2
25 g
≥98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Iron(III) bromide, anhydrous

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Iron bromide

SDS Documents

Documents

Certificates
Special Offers

Special Offers