missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) chloride hexahydrate, 97+%, ACS reagent

Product Code. 15119293 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic Bottle
Plastic drum
Unit Size:
10kg
2.5kg
500g
5g
This item is not returnable. View return policy

Product Code. 15119293

Brand: Thermo Scientific Acros 423700100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10025-77-1 | Cl3FeH12O6 | 270.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Ferric chloride hexahydrate
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Boiling Point 280.0°C to 285.0°C
Melting Point 37.0°C
Color White
Physical Form Crystalline Powder
Quantity 10 kg
Assay Percent Range 97.0 to 102% (Iodometry)
Linear Formula FeCl3·6H2O
Fieser 01,390; 02,199; 03,145; 04,236; 05,307; 06,259; 07,153; 08,228; 09,222; 15,158
Merck Index 15,4046
Solubility Information Solubility in water: 920g/L (20°C). Other solubilities: readily soluble in alcohol, acetone and ether
Formula Weight 270.29
Percent Purity ≥97%
Grade ACS Reagent
Packaging Plastic drum
Chemical Name or Material Iron(III) chloride hexahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(III) chloride hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P313-Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.