missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) chloride hexahydrate, 97%

Product Code. 11403347
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11403347 2500 g 2500g
11493337 500 g 500g
11413347 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403347 Supplier Thermo Scientific Alfa Aesar Supplier No. A16231.0E
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10025-77-1 | Cl3FeH12O6 | 270.29 g/mol

Iron(III) chloride hexahydrate is used to prepare organically templated iron phosphates. It is used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin process for the production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards. It plays an important role in photoengraving and photography as an etching agent. It is useful as a mordant in dyeing and printing textiles. It is useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(III) chloride hexahydrate is used to prepare organically templated iron phosphates. It is used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin process for the production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards. It plays an important role in photoengraving and photography as an etching agent. It is useful as a mordant in dyeing and printing textiles. It is useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids.

Solubility
Soluble in water, alcohol, glycerol, methanol, ether and acetone. Insoluble in ethyl acetate.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, sodium and sodium oxides and potassium.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Ferric chloride hexahydrate
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Spezifikation

Boiling Point 280°C to 285°C
Melting Point 37°C
pH 1.8
Quantity 2500 g
Odor Odorless
Sensitivity Hygroscopic
Density 1.82
Linear Formula FeCl3·6H2O
UN Number UN3260
Merck Index 14,4019
Solubility Information Soluble in water,alcohol,glycerol,methanol,ether and acetone. Insoluble in ethyl acetate.
Formula Weight 270.30 (162.21 Anhydrous)
Percent Purity 97%
Chemical Name or Material Iron(III) chloride hexahydrate
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • Iron(III) chloride hexahydrate
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Gefahrenhinweis
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Ergänzende Informationen
  • MIXTURE LIST-Contain: Ferric chloride hexahydrate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.