missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Chloride Hexahydrate, ≥98%, Reagent Grade, Honeywell™

Product Code. 15657860 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Packaging:
Poly bottle
Unit Size:
100g
1kg
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15657860 100 g Poly bottle 100g
15677860 250 g Poly bottle 250g
15687860 500 g Poly bottle 500g
15667860 1 kg Poly bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15657860 Supplier Honeywell Research Chemicals Supplier No. F2877100G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, chunks,≥98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Boiling Point 280°C to 285°C
Melting Point 37°C
Color Brown
pH 1.8 (at 10.00g/L 25°C)
Physical Form Solid
Quantity 100 g
Linear Formula FeCl3 · 6H2O
UN Number UN3260
Formula Weight 270.3g/mol
Percent Purity ≥98%
Odor Weak
Packaging Poly bottle
Vapor Pressure 1mmHg (194°C)
Density 2.8g/cu.cm at 20°C
Chemical Name or Material Iron(III) chloride hexahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.