missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) chloride hexahydrate, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 50 g |
| Linear Formula | FeCl3 · 6H2O |
| UN Number | UN3260 |
| Formula Weight | 270.3g/mol |
| Percent Purity | 98 to 102% (RT) |
| Vapor Pressure | 1 mmHg (194°C) |
| Chemical Name or Material | Iron(III) chloride hexahydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction