missing translation for 'onlineSavingsMsg'
Learn More

Iron(Iii) Chloride, Honeywell

Product Code. 15631510 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
2.5 kg
20 kg
Unit Size:
100g
1kg
2.5kg
20kg
5g
Product Code. Quantity unitSize
15631510 5 g 5g
15691500 100 g 100g
15601510 1 kg 1kg
15611510 2.5 kg 2.5kg
15621510 20 kg 20kg
5 options
This item is not returnable. View return policy

Product Code. 15631510

Brand: Honeywell Research Chemicals 1577405G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7705-08-0
Molecular Formula Cl3Fe
Molecular Weight (g/mol) 162.20
MDL Number MFCD00011005
InChI Key RBTARNINKXHZNM-UHFFFAOYSA-K
IUPAC Name iron(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Quantity 5 g
Linear Formula FeCl3
UN Number 1773
Formula Weight 162.2g/mol
Percent Purity 97%
Grade Reagent
Vapor Pressure 1mmHg (194°C)
Density Vapor Density: 5.61 (vs air)
Chemical Name or Material Iron(Iii) Chloride
compliance-icons compliance-icons
Product Identifier
  • Iron(III) chloride
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.