missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) chloride solution, Purum, 45% FeCl3 basis, Honeywell Fluka™

Product Code. 15641120 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
Unit Size:
2.5 L
Pack of 4
This item is not returnable. View return policy

Product Code. 15641120

Brand: Honeywell-Fluka 123222.5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum, 45% FeCl3 basis

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7705-08-0
Molecular Formula Cl3Fe
Molecular Weight (g/mol) 162.20
MDL Number MFCD00011005
InChI Key RBTARNINKXHZNM-UHFFFAOYSA-K
IUPAC Name iron(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Quantity 2.5 L
Linear Formula FeCl3
UN Number 2582
Formula Weight 162.2g/mol
Grade purum
Concentration or Composition (by Analyte or Components) 45% FeCl3 basis
Chemical Name or Material Iron(Iii) Chloride Solution
compliance-icons compliance-icons
Product Identifier
  • Iron(III) chloride solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Iron trichloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.