missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Chloride, Technical, Solution, about 60% (w/v), FeCl3, Fisher Chemical

Product Code. 10417260 Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Unit Size:
2.5 L
This item is not returnable. View return policy

Product Code. 10417260

Brand: Fisher Chemical I/1045/PB17

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7705-08-0
Color Brown
Physical Form Liquid
Quantity 2.5 L
Molecular Formula Cl3Fe
MDL Number MFCD00011005
InChI Key RBTARNINKXHZNM-UHFFFAOYSA-K
SMILES [Cl-].[Cl-].[Cl-].[Fe+3]
IUPAC Name iron(3+) trichloride
Molecular Weight (g/mol) 162.20
Packaging HDPE plastic bottle
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron (III) Chloride Solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.