missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) Chloride, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Iron(III) Chloride |
| CAS | 7705-08-0 |
| Molecular Formula | Cl3 Fe |
| Quantity | 10 g |
| Synonym | Bellair green plus, Clarfer, Clayfec, Danube clear classic, Donau klar classic, FC-Pix, Ferric chloride, Ferric trichloride, Ferrisol Cl 130, Flores martis, Iron chloride, Iron perchloride, Iron trichloride, NSC 135798, NSC 51150, Riedel 12322, Trifloc, ViscoStat Plus |
| InChI Formula | InChI=1S/3ClH.Fe/h3*1H;/q;;;+3/p-3 |
| SMILES | Cl[Fe](Cl)Cl |
| IUPAC Name | trichloroiron |
| Molecular Weight (g/mol) | 162.204 |
| Formula Weight | 160.8415 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction