missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) citrate hydrate, 98%

Product Code. 10164740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10164740 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10164740 Supplier Thermo Scientific Acros Supplier No. 211850010
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 334024-15-6 | C6H5FeO7 | 244.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 334024-15-6
Color White
Infrared Spectrum Authentic
Assay Percent Range 18 to 22% (Fe) (Iodometry)
Packaging Plastic bottle
Molecular Formula C6H5FeO7
MDL Number MFCD00149615
Quantity 1 kg
Merck Index 14, 4021
Synonym ferric citrate monohydrate, iron iii citrate hydrate, unii-334s8t6lke, ferric citrate hydrate, ferric citrate usan, iron iii citrate tribasic monohydrate, citric acid iron iii salt, iron cento citrate hydrate, ksc534c4h, iron 3+ citrate hydrate
Solubility Information Solubility in water: 5g/L (25°C). Other solubilities: insoluble in ethanol, soluble in hno3
InChI Key NPFOYSMITVOQOS-UHFFFAOYSA-K
SMILES [Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylate;iron(3+);hydrate
Molecular Weight (g/mol) 244.94
PubChem CID 22178220
Formula Weight 262.96
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Iron(III) citrate hydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.