missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) citrate hydrate, Fe(III) 16.5-20%; Fe(II) max 5%

Product Code. 11494003
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494003 250 g 250g
11484003 50 g 50g
11474003 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494003 Supplier Thermo Scientific Alfa Aesar Supplier No. A10276.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 334024-15-6 | C6H5FeO7 | 244.94 g/mol

Provides a source of iron used in cell culture applications. Provides iron in a less toxic form than free iron salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Provides a source of iron used in cell culture applications. Provides iron in a less toxic form than free iron salts.

Solubility
Soluble in water.

Notes
Light sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 334024-15-6
Molecular Formula C6H5FeO7
Molecular Weight (g/mol) 244.94
MDL Number MFCD00149615
InChI Key NPFOYSMITVOQOS-UHFFFAOYSA-K
Synonym ferric citrate monohydrate, iron iii citrate hydrate, unii-334s8t6lke, ferric citrate hydrate, ferric citrate usan, iron iii citrate tribasic monohydrate, citric acid iron iii salt, iron cento citrate hydrate, ksc534c4h, iron 3+ citrate hydrate
PubChem CID 22178220
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylate;iron(3+);hydrate
SMILES [Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Quantity 250 g
Solubility Information Soluble in water.
Formula Weight 244.94 (Anhydrous)
Concentration or Composition (by Analyte or Components) Fe(III) 16.5 to 20%; Fe(II) max 5%
Sensitivity Light sensitive
Chemical Name or Material Iron (III) citrate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.