missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) ethoxide, soluble polymeric mixture, Thermo Scientific™

Product Code. 15472467
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15472467 2 g 2g
15482467 10 g 10g
15492467 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15472467 Supplier Thermo Scientific Alfa Aesar Supplier No. 045861.04
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5058-42-4
Molecular Formula C6H15FeO3
Molecular Weight (g/mol) 191.028
MDL Number MFCD00078028
InChI Key JTPUGUWXHGEEHW-UHFFFAOYSA-N
Synonym iron iii ethoxide, iron 3+ ethanolate, triethoxyiron iii, ethanolate; iron 3+, acmc-1alnr, iron 3+ tris ethoxide, iron 3+ ion tris ethoxide, iron iii ethoxide trace metals basis 500mg
PubChem CID 6452222
IUPAC Name ethanolate;iron(3+)
SMILES CC[O-].CC[O-].CC[O-].[Fe+3]

Specifications

Color Brown
Boiling Point (decomposition)
Quantity 2 g
UN Number UN2926
Solubility Information Soluble in hexane and ethanol
Physical Form Solid
Chemical Name or Material Iron (III) ethoxide, Soluble polymeric mixture
compliance-icons compliance-icons
Product Identifier
  • Iron(III) ethoxide, soluble polymeric mixture
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H228-Flammable solid.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.