missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) hydroxide, 99+%

Product Code. 15473127
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15473127 250 g 250g
15463127 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473127 Supplier Thermo Scientific Alfa Aesar Supplier No. A16267.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 20344-49-4 | FeHO2 | 88.851 g/mol

Iron(III) hydroxide, alpha-phase, is used as a catalyst for proteomics research. It is also used as a pharamaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(III) hydroxide, alpha-phase, is used as a catalyst for proteomics research. It is also used as a pharamaceutical intermediate.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20344-49-4
Molecular Formula FeHO2
Molecular Weight (g/mol) 88.851
MDL Number MFCD00064782
InChI Key AEIXRCIKZIZYPM-UHFFFAOYSA-M
Synonym hydroxy oxo iron, goethite, lepidocrocite, iron hydroxide oxide, goethite fe oh o, iron hydroxide oxide fe oh o, iron iii oxide-hydroxide, ferric acid, iron pigment yellow, ferric hydroxide oxide
PubChem CID 91502
IUPAC Name hydroxy(oxo)iron
SMILES O[Fe]=O

Specifications

Quantity 250 g
Linear Formula FeO(OH)
Merck Index 14,4025
Solubility Information It is insoluble in water.
Formula Weight 88.85
Percent Purity ≥99%
Odor Odorless
Density 4.1
Chemical Name or Material Iron(III) hydroxide, alpha-phase

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.