Learn More
Iron(III) hydroxide, alpha-phase
Description
Iron(III) hydroxide is used in purifying water and as an absorbent in chemical processes. It is also used as a pigment and as a catalyst. Further, it is used in some tattoo ink and in aquarium water treatment as a phosphate binder.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Brown |
| Physical Form | Powder |
| Quantity | 5 g |
| Solubility Information | Soluble in mineral acids. Insoluble in water and ethanol. |
| Formula Weight | 88.85 |
| Appearance | Brown powder |
| Density | 3.4 to 3.9 g/cm3 |
| Chemical Name or Material | Iron(III) hydroxide |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.