missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) hydroxide, alpha-phase

Product Code. 11373587
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373587 25 g 25g
11363587 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373587 Supplier Thermo Scientific Alfa Aesar Supplier No. 019496.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

For Use in Purifying Water, As Absorbent in Chemical Processes, As a Pigment | CAS: 20344-49-4 | FeHO2 | 88.851 g/mol

Iron(III) hydroxide is used in purifying water and as an absorbent in chemical processes. It is also used as a pigment and as a catalyst. Further, it is used in some tattoo ink and in aquarium water treatment as a phosphate binder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20344-49-4
Molecular Formula FeHO2
Molecular Weight (g/mol) 88.851
MDL Number MFCD00064782
InChI Key AEIXRCIKZIZYPM-UHFFFAOYSA-M
Synonym hydroxy oxo iron, goethite, lepidocrocite, iron hydroxide oxide, goethite fe oh o, iron hydroxide oxide fe oh o, iron iii oxide-hydroxide, ferric acid, iron pigment yellow, ferric hydroxide oxide
PubChem CID 91502
IUPAC Name hydroxy(oxo)iron
SMILES O[Fe]=O

Specifications

Color Brown
Physical Form Powder
Quantity 25 g
Solubility Information Soluble in mineral acids. Insoluble in water and ethanol.
Formula Weight 88.85
Appearance Brown powder
Density 3.4-3.9
Chemical Name or Material Iron(III) hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.