missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) isopropoxide, 98%

Product Code. 11396289
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11396289 1 g 1g
11306299 5 g 5g
11316299 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11396289

Brand: Thermo Scientific Alfa Aesar 044873.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14995-22-3 | C9H21FeO3 | 233.109 g/mol

Iron(III) isopropoxide is applied in the field of nano-materials and thin films, engineering and environmental field. It is employed in catalysis and synthesis, energy conversion and storage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14995-22-3
Molecular Formula C9H21FeO3
Molecular Weight (g/mol) 233.109
MDL Number MFCD00070436
InChI Key QUHDSMAREWXWFM-UHFFFAOYSA-N
Synonym iron 3+ ;propan-2-olate, iron iii i-propoxide, triisopropoxyiron iii, 2-propanol, iron 3+ salt 9ci, iron 3+ ion tris propan-2-olate, iron 3+ tris propan-2-olate, iron iii i-propoxide 99.9+%-fe
PubChem CID 11961763
IUPAC Name iron(3+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[Fe+3]

Specifications

Color Gray
Packaging Ampule
Linear Formula Fe[OCH(CH3)2]3
Quantity 1 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 233.11
Percent Purity 98%
Physical Form Solid Mass
Chemical Name or Material Iron (III) isopropoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(III) isopropoxide
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.