missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) nitrate nonahydrate, 98+% (metals basis)

Product Code. 11396638
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
Unit Size:
100g
1kg
5kg
Product Code. Quantity unitSize
11396638 100 g 100g
11306648 1 kg 1kg
11316648 5 kg 5kg
3 options
This item is not returnable. View return policy

Product Code. 11396638

Brand: Thermo Scientific Alfa Aesar 012226.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7782-61-8 | FeH18N3O18 | 403.99 g/mol

It is used as catalyst for the synthesis of sodium amide from sodium and ammonia, and for etching of silver and silver alloys. Ferric nitrate is used in the synthesis of organically templated iron phosphates, and acts as an efficient catalyst in enamination of beta dicarbonyl compounds to beta enaminones. Ferric nitrate impregnated clays are useful as oxidizing agents. Clayfen, produced from ferric nitrate and montmorillonite, is used for the oxidation of thiols to disulfide, and alcohols to aldehydes. Ferric nitrate is useful for regio- and stereo-selective nitration of various aromatic, aliphatic, and heteroaromatic olefins. This reaction provides nitroolefins with excellent E-selectivity. With Keggin-type heteropoly acids, it selectively oxidizes various hydroxy groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-61-8
Molecular Formula FeH18N3O18
Molecular Weight (g/mol) 403.99
MDL Number MFCD00149708
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
Synonym ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
PubChem CID 16211566
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 125°C (decomposition)
Melting Point 47°C
Color Purple
Physical Form Crystalline
Quantity 100 g
Assay Percent Range >98% (metals basis)
Solubility Information Soluble in water,alcohol and acetone.
IUPAC Name iron(3+);trinitrate;nonahydrate
Formula Weight 403.99 (241.86 Anhydrous)
Odor Acidic
Sensitivity Moisture sensitive
Appearance Pale purple crystalline
Density 1.684 g/cm3
Chemical Name or Material Iron(III) nitrate nonahydrate, Contains excess HNO3
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(III) nitrate nonahydrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.