missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Nitrate Nonahydrate, TRC

Product Code. 30391361
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30391361 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30391361

Brand: Toronto Research Chemicals TRCI77521510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iron(III) Nitrate Nonahydrate
CAS 7782-61-8
Molecular Formula Fe(NO3)3 . 9H2O
Quantity 10 g
InChI Formula InChI=1S/Fe.3NO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;;;;9*1H2/q+3;3*-1;;;;;;;;;
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+](=O)[O-].[O-][N+](=O)[O-].[O-][N+](=O)[O-]
IUPAC Name iron(3+);trinitrate;nonahydrate
Molecular Weight (g/mol) 404
Formula Weight 403.9935
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.