missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) oxide, 98% (metals basis), Thermo Scientific Chemicals

Product Code. 11361769
Change view
Click to view available options
Quantity:
1 kg
25 kg
Unit Size:
1kg
25kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11361769 25 kg 25kg
11341769 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361769 Supplier Thermo Scientific Alfa Aesar Supplier No. 012375.C4
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

It has many applications, mainly used in iron industry, pigment, as photocatalyst, medicine and polishing jewels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O

Specifications

Melting Point 1565°C
Color Red to Brown
Physical Form Powder
Quantity 25 kg
Assay Percent Range 98%
Solubility Information Insoluble in water,Soluble in diluted acids,sugar solution and slightly soluble in aq. tartaric acid,citric acid and acetic acid.
Formula Weight 159.69
Appearance Red-brown powder
Density 5.24
Chemical Name or Material Iron(III) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.