missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Oxide, 99.8%, Trace metals basis, Solstice

Product Code. 15654160 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Poly bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15654160 50 g Poly bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15654160 Supplier Honeywell Research Chemicals Supplier No. 34300550G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Pieces, 3-12 mm, 99.8% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Spezifikation

Melting Point 1565°C
CAS 1309-37-1
Color Red
pH 4 to 8 (at 50.00g/L)
Physical Form Solid
Quantity 50 g
Molecular Formula Fe2O3
MDL Number MFCD00011008
UN Number NONH for all modes of transport
Solubility Information Insoluble in water
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
SMILES [Fe][Fe](=O)(=O)=O
IUPAC Name trioxodiiron
Molecular Weight (g/mol) 159.69
Formula Weight 159.69g/mol
Percent Purity 99.8% (trace metals basis)
Odor Odorless
Packaging Poly bottle
Density ∽5g/cu.cm at 20°C
Chemical Name or Material Iron(III) oxide
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.