missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) oxide, 99.999%, (trace metal basis), -100 mesh

Product Code. 10256250 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10256250 10 g Glass bottle 10g
10072550 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10256250 Supplier Thermo Scientific Acros Supplier No. 193260100
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1309-37-1 | Fe2O3 | 159.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
Synonym Ferric oxide
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O

Specifications

Melting Point 1538.0°C
Color Brown to Brown-Red or Rust
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.999%
Linear Formula Fe2O3
Fieser 17,158
Merck Index 15,4055
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids, soluble in alcohol and ether
Formula Weight 159.69
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 5.24
Density 5.2400g/mL
Chemical Name or Material Iron(III) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.