missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) oxide, Purified, ≥95%, Solstice

Product Code. 16685733
Change view
Click to view available options
10 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16685733 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16685733 Supplier Fluka Supplier No. 1234210KG
 Restricted Item
Estimated Shipment: 15-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 1,565°C
CAS 1309-37-1
Color Red
CAS Max % 1
pH 4 to 8
Quantity 10 kg
Molecular Formula Fe2O3
Linear Formula Fe2O3
MDL Number MFCD00011008
Synonym Ferric oxide
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
SMILES [Fe][Fe](=O)(=O)=O
IUPAC Name trioxodiiron
Molecular Weight (g/mol) 159.69
Grade Synthesis
Density 5 g/cm3 (20°C)
Chemical Name or Material Iron(III) oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.