missing translation for 'onlineSavingsMsg'
Learn More

Iron(Iii) Oxide, purified, Solstice

Product Code. 15641130
Change view
Click to view available options
Quantity:
250 g
1 kg
6 x 250 g
6 x 1 kg
Unit Size:
1kg
250g
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15641130 6 x 250 g Pack of 6
15621130 250 g 250g
15611130 1 kg 1kg
15631130 6 x 1 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15641130 Supplier Honeywell-Fluka Supplier No. 123426X250G
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Purified,≥95%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O

Specifications

Quantity 6 x 250 g
UN Number NONH for all modes of transport
Formula Weight 159.69
Percent Purity ≥95%
Grade purified
Chemical Name or Material Iron(Iii) Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.