missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Oxide, TRC

Product Code. 30406482
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406482 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406482 Supplier Toronto Research Chemicals Supplier No. TRCI7752102.5G
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iron (III) Oxide
CAS 1309-37-1
Quantity 2.5 g
Molecular Formula Fe2 O3
Synonym Iron Oxide Pigment Red, Iron oxide (Fe2O3), α-Ferric oxide, α-Iron oxide, γ-Ferric oxide, γ-Iron oxide (Fe2O3), γ-MRD, γ-MYD, Iron Oxide Red, Iron Oxide Red 110M, Iron Oxide Red 130A, Iron Oxide Red 130M, Iron Oxide Red N 135, Iron Oxide Red Transparent 288VN, Iron Oxides CI 77491, Iron Red 2597, Iron Red 3602, Iron Red 4597, Iron Red 5597, Iron Red 6597, Iron Red TP 303, Iron minium, Iron oxide, Iron oxide (Fe2O3) nanoparticles, Iron oxide (FeO1.5), Iron red, Iron sesquioxide, Iron sesquioxide (Fe2O3), Iron trioxide, Iron(3+) oxide, Iron(III) oxide, Pigment Red 101
InChI Formula InChI=1S/2Fe.3O
SMILES O=[Fe]O[Fe]=O
IUPAC Name oxo(oxoferriooxy)iron
Molecular Weight (g/mol) 159.69
Formula Weight 159.8546
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.