missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) sulfate pentahydrate, 97%

Product Code. 10460863 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
5 kg
Packaging:
Glass Bottle
Plastic Drum
Plastic bottle
Unit Size:
100g
500g
5g
5kg
This item is not returnable. View return policy

Product Code. 10460863

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 142906-29-4 | Fe2O12S3·5H2O | 489.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142906-29-4
Molecular Formula Fe2O12S3·5H2O
Molecular Weight (g/mol) 489.96
MDL Number MFCD00149714
InChI Key YHGPYBQVSJBGHH-UHFFFAOYSA-H
Synonym unii-3hws7hf5xd, 3hws7hf5xd, iron iii sulfate pentahydrate, sulfuric acid, iron 3+ salt 3:2 , pentahydrate 9ci, acmc-20n1we, ferric sulfate pentahydrate, iron iii sulphate pentahydrate, 2fe.3so4.5h2o, sulfuric acid, iron 3+ salt 3:2 , pentahydrate
PubChem CID 23443659
IUPAC Name iron(3+);trisulfate;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3]

Specifications

Color Beige-Yellow to White
Physical Form Crystalline or Granular Powder
Quantity 500 g
Assay Percent Range 97%
Linear Formula Fe2(SO4)3·5H2O
Solubility Information Solubility in water: soluble
Formula Weight 489.96
Percent Purity 97%
Packaging Plastic bottle
Chemical Name or Material Iron(III) sulfate pentahydrate
compliance-icons compliance-icons
Product Identifier
  • Iron(III)sulfate pentahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.