missing translation for 'onlineSavingsMsg'
Learn More

Iron Oxide, Pure, Magnetic, Precipitated, Fisher Chemical

Product Code. 10385990 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10385990 500 g Glass Jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10385990 Supplier Fisher Chemical Supplier No. I/1100/53
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

Melting Point 1588.9°C
CAS 1317-61-9
Color Black
pH 4 to 8
Physical Form Solid
Quantity 500 g
Molecular Formula Fe3O4
MDL Number MFCD00011010
InChI Key WTFXARWRTYJXII-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[O--].[Fe++].[Fe+3].[Fe+3]
IUPAC Name diiron(3+) λ²-iron(2+) tetraoxidandiide
Molecular Weight (g/mol) 231.53
Formula Weight 231.5386g/mol
Packaging Glass Jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.