missing translation for 'onlineSavingsMsg'
Learn More

Iron phthalocyanine

Product Code. 10403741 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10403741 25 g Glass bottle 25g
10341111 5 g Glass bottle 5g
11985861 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10403741 Supplier Thermo Scientific Acros Supplier No. 412610250
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 132-16-1 | C32H16FeN8 | 568.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-16-1
Molecular Formula C32H16FeN8
Molecular Weight (g/mol) 568.38
MDL Number MFCD00015953
InChI Key MIINHRNQLVVCEW-UHFFFAOYSA-N
Synonym iron phthalocyanine, iron ii phthalocyanine, ferrous phthalocyanine, phthalocyanine iron ii, fepc, 29h,31h-phthalocyaninato 2--n29,n30,n31,n32 iron, iron, 29h,31h-phthalocyaninato 2--n29,n39,n31,n32-, sp-4-1, iron, 29h,31h-phthalocyaninato 2--kappan29,kappan30,kappan31,kappan32-, sp-4-1
PubChem CID 2735065
IUPAC Name λ²-iron(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
SMILES [Fe++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31

Specifications

Color Black or Green-Purple
Physical Form Powder
Quantity 25 g
Solubility Information Solubility in water: .. Other solubilities: soluble in conc. sulfuric acid
Formula Weight 568.38
Percent Purity 70 to 90% (typically)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Iron phthalocyanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.