missing translation for 'onlineSavingsMsg'
Learn More

Iron powder, spherical, APS 6-10 micron, reduced, 99.5%, Thermo Scientific Chemicals

Product Code. 15445925
Change view
Click to view available options
Quantity:
25 g
250 g
500 g
2 x 500 g
Unit Size:
250g
25g
500g
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15445925 25 g 25g
15455925 250 g 250g
15465925 500 g 500g
15475925 2 x 500 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15445925 Supplier Thermo Scientific Alfa Aesar Supplier No. 010214.14
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Apparent density 2.0-3.0g/cm3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Color Gray
Appearance Gray powder
Density 7.87g/cm3 at 20°C
Quantity 25 g
Assay Percent Range 99.5% (Metals basis)
Form Spherical Powder
Chemical Name or Material Iron, Approximately 98% spherical, 2% fragments

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.