missing translation for 'onlineSavingsMsg'
Learn More

Iron sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)

Product Code. 15414795
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414795 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15414795 Supplier Thermo Scientific Alfa Aesar Supplier No. 040854.KS
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7439-89-6 | Fe | 55.85 g/mol

Magnetron sputtering source

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Ea.
CAS 7439-89-6
Molecular Formula Fe
MDL Number MFCD00010999
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES [Fe]
IUPAC Name iron
Molecular Weight (g/mol) 55.85
PubChem CID 23925
ChEBI CHEBI:82664
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Iron sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.