missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Iron Test Kits

Product Code. 11795783
Change view
Click to view available options
Range:
1 to 50 mg/L
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
11795783 1 to 50 mg/L Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11795783 Supplier WTW™ Supplier No. 250361
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Convenient, cost effective, precise; these Iron (Fe) cell test kit product types are ideal for testing waste water, drinking water, and foodstuffs.

  • For use with PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral photometers
  • For waste water, drinking water, foodstuffs
  • Iron (Fe) cell test kit product type
  • Triazine method
  • Photometric method
  • Convenient, cost effective, precise
TRUSTED_SUSTAINABILITY

Specifications

Analytes Triazine
For Use With (Application) For waste water, drinking water, foodstuffs
For Use With (Equipment) PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral photometers
Includes Certificates
No. of Tests 25
Range 1 to 50 mg/L
Sufficient For 25 Tests
Test Method Photometric
Type Iron (Fe) cell test kit
compliance-icons
Product Identifier
  • Tin Cell Test Method: photometric 0.10 - 2.50 mg/l Sn Spectroquant
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: hydrochloric acid, N-Cetyl-N'N'N-trimethylammonium bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.