missing translation for 'onlineSavingsMsg'
Learn More

Iron wire, 0.25mm (0.01in) dia, Puratronic™, 99.99% (metals basis)

Product Code. 11331488
Change view
Click to view available options
Quantity:
1 m
5 m
25 m
Unit Size:
1m
25m
5m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11331488 1 m 1m
11321488 5 m 5m
11311488 25 m 25m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11331488 Supplier Thermo Scientific Alfa Aesar Supplier No. 010937.G1
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Form: approx. 0.39g/m | CAS: 7439-89-6 | Fe | 55.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Color Gray
Appearance Gray wire
Density 7.87g/cm3 at 20°C
Quantity 1 m
Assay Percent Range 99.995% (Metals basis)
Form Wire
Chemical Name or Material Iron

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.