missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Iron Wire Brushes

Product Code. 10204131
Change view
Click to view available options
Length (Metric):
270 mm
340 mm
380 mm
420 mm
470 mm
Length (Metric) Brush:
100 mm
120 mm
130 mm
140 mm
170 mm
For Use With (Application):
Flasks
Test tube
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Length (Metric) Brush For Use With (Application) Unit Size
10204131 270 mm 100 mm Test tube Each
10429392 340 mm 120 mm Flasks Each
10080781 380 mm 130 mm Flasks Each
10631732 420 mm 140 mm Flasks Each
10586371 470 mm 170 mm Flasks Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10204131 Supplier Bochem™ Supplier No. 12310
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Made with natural bristles. Bochem™ Iron Wire Brushes are used for test tubes and flasks.

  • Iron wire, zincked
  • Natural bristles
  • For test tubes and flasks

Specifications

Product Type Test Tube Brush
For Use With (Application) Test tube
Length (Metric) 270 mm
Length (Metric) Brush 100 mm
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.