missing translation for 'onlineSavingsMsg'
Learn More

Iron yttrium oxide, 99.9% (REO)

Product Code. 11348288
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11348288

Brand: Thermo Scientific Alfa Aesar 039488.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12063-56-8 | Fe5O12Y3 | 737.931 g/mol

Iron yttrium oxide is mainly used in superconductors. It is also used in filters, transmitters, transducers, solid state lasers in faraday rotators and in non-linear optics. It finds application in polymers, textiles, fuel cell layers, composites and solar energy materials, bio nano materials and bio sensors. Further, it is used in the preparation of fuel cells and solar energy materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12063-56-8
Molecular Formula Fe5O12Y3
Molecular Weight (g/mol) 737.931
MDL Number MFCD00210618
InChI Key UVXIKKWNYGPENJ-UHFFFAOYSA-N
Synonym iron yttrium oxide, yttrium iron garnet, yttrium ferrite garnet, bis ferric oxide ; oxo oxoyttrio oxy iron; oxo oxoyttrio oxy yttrium, yttrium iron oxide, nanopowder, <100 nm particle size bet trace metals basis
PubChem CID 16217670
IUPAC Name oxo(oxoferriooxy)iron;oxo(oxoferriooxy)yttrium;oxo(oxoyttriooxy)yttrium
SMILES O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O.O=[Fe]O[Y]=O.O=[Y]O[Y]=O

Specifications

Color Green
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9% (REO)
Solubility Information Insoluble in water.
Formula Weight 737.94
Odor Odorless
Appearance Green powder
Chemical Name or Material Iron yttrium oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.