Learn More
Chemsplash Irradiated Zip Labcoat PB(6B) Elastic Cuff
Product Code. 30058406
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30058406 2X-Large Pack of 50
30058299 Small Pack of 50
30058208 Medium Pack of 50
30058234 Large Pack of 50
30058374 X-Large Pack of 50
30058289 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30058406 Supplier Chemsplash Supplier No. 2635.400.008
  • ONLINE EXCLUSIVE PRICE
    298.20 €
    251.24 € / Pack of 50
    Save 46.96 €
    15% Off
Estimated Shipment: 26-07-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Supplied Gamma Sterile Irradiated and double bagged. This lab coat is a 2/3 length coat with a collar and zip closure and elasticated wrists.
Chemsplash Sterile Lab Coat
  • Chemsplash 55GSM Type 5B and 6B PB Fabric
  • GAMMA Irradiated and Double Bagged
  • Zip Fasten Front with Adhesive Flap Cover
  • Elasticated Cuffs
  • Fold Over Collar
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting
  • Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9
Description: Supplied GAMMA Sterile Irradiated and double bagged, the Chemsplash Lab Coat is made from Chemsplash fabric, which is soft and microporous. It is strong and resistant to chemical splash and dust. This lab coat is a 2/3 length coat with a collar and zip closure and elasticated wrists. It is anti-static to meet with EN1149-5:2008, lint-free and suitable for clean room applications. Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9. Chemsplash accessories can be used to protect the wearer from the environment or the environment from the wearer. The range has been designed to allow for maximum comfort and wearability.
Specifications
Product Type Lab Coat
Size 2X-Large
Gender Unisex
Material Microporous Composite Fabric
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Single-use
Sterility Sterile
Resistance Liquids
No. of Pockets None
Latex Free Yes
Autoclavable Autoclavable
Cleanroom Class Class 100-100,000 (ISO 5-8)