Learn More
Chemsplash Irradiated Zip Labcoat PB(6B) Thumb Loops, Elastic Cuff
Product Code. 30058365
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30058365 Large Pack of 50
30058345 Small Pack of 50
30058288 Medium Pack of 50
30058188 X-Large Pack of 50
30058285 2X-Large Pack of 50
30058156 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30058365 Supplier Chemsplash Supplier No. 2637.400.006
  • ONLINE EXCLUSIVE PRICE
    509.00 €
    457.17 € / Pack of 50
    Save 51.83 €
    10% Off
Estimated Shipment: 24-05-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Supplied gamma Sterile Irradiated and double bagged. This lab coat is a 2/3 length coat with a collar and zip closure and elasticated wrists with thumb loops.
Chemsplash Lab Coat
  • Chemsplash 55GSM Type PB 6B Fabric
  • GAMMA Irradiated and Double Bagged
  • Zip Fasten Front with Adhesive Flap Cover
  • Elasticated Cuffs with Thumb Loops
  • Fold Over Collar
  • Silicone and Latex Free, PFAS Free
  • Anti-Static Material
  • Non-Linting
  • Conforms to ISO 14644-1:2015 Cleanroom Class 5 - 9
Description: The Chemsplash Lab Coat is supplied GAMMA Sterile Irradiated and double bagged. Made from Chemsplash fabric, it is soft and microporous, offering strength and resistance to chemical splash and dust. This lab coat features a 2/3 length design with a collar and zip closure, as well as elasticated wrists with thumb loops for added convenience. It is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforms to ISO 14644-1:2015 Cleanroom Class 5 - 9. Chemsplash accessories protect the wearer from the environment or the environment from the wearer, designed for maximum comfort and wearability.
Specifications
Product Type Cleanroom Coveralls
Size Large
Gender Unisex
Material Microporous Composite Fabric
Color White
Disposable Single-use
Waist Style Loose
Ankle Style Open
Cuff Style Elastic, Thumb Loop
Neck Style Collared
Hood Type Unhooded
Closure Type Ties
Seam Type Stitched
Sterility Sterile
Resistance Liquid
Latex Free Yes
No. of Pockets None
Cleanroom Class Class 100-100,000 (ISO 5-8)