missing translation for 'onlineSavingsMsg'
Learn More

Isethionic acid, potassium salt, 98%, Thermo Scientific™

Product Code. 10557362 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
25g; Glass bottle
Product Code.
10557362 25g; Glass bottle
1 options
This item is not returnable. View return policy

Product Code. 10557362

Brand: Thermo Scientific Acros 412630250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1561-99-5
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C2H5KO4S
Linear Formula HOCH2CH2SO3K
Quantity 25g
Synonym potassium isethionate, potassium 2-hydroxyethanesulfonate, potassium 2-hydroxyethanesulphonate, ethanesulfonic acid, 2-hydroxy-, monopotassium salt, isethionic acid potassium salt, 2-hydroxyethanesulfonic acid potassium salt, phes, ethanesulfonic acid, 2-hydroxy-, potassium salt 1:1, isethionic acid, potassium salt, 2-hydroxy-ethanesulfonicacimonopotassiumsalt
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol
InChI Key DMNPUBKRNRRYMC-UHFFFAOYSA-M
SMILES C(CS(=O)(=O)[O-])O.[K+]
IUPAC Name potassium;2-hydroxyethanesulfonate
Molecular Weight (g/mol) 164.216
PubChem CID 23689117
Formula Weight 164.22
Percent Purity 98%
Chemical Name or Material Isethionic acid, potassium salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.